Evaporating bowls, with spout
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
796 798
NICK796EA 46.7 EUR
NICK796 NICK798
Evaporating bowls, with spout
Reservoirs/Schalen/Potten Uitdampschalen
Stainless steel.
  • Corrosion resistant
  • Excellent thermal resistance
  • Smooth surfaces for easy cleaning
Multipurpose bowls ideal for a wide variety of uses.