Test tubes,with screw thread
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
716150 716100 713100 716125 720150 617145
DELT716150EA 444 EUR
DELT716150 DELT716100 DELT713100 DELT716125 DELT720150 DELT617145
Test tubes,with screw thread
Buisjes Reageerbuisjes
These screw threaded glass tubes are made of borosilicate or soda glass.
  • Polypropylene caps are designed for the round bottom tubes
  • Aluminium caps are designed for flat bottom tubes
  • Resistant to 4000 min⁻¹
Leveringsinformatie: Supplied in small quantities per case for a more convenient use in laboratory. Tubes are supplied without caps.