Microtube racks, with lid
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-020R M-020V M-020AM M-020N U-9950CV U-7780N U-7750N U-8845A U-7796V
DELTM-020REA 27.9 EUR
DELTM-020R DELTM-020V DELTM-020AM DELTM-020N DELTU-9950CV DELTU-7780N DELTU-7750N DELTU-8845A DELTU-7796V
Microtube racks, with lid
Rekken Tube Racks
PP, square with translucent lid.
  • Range available for variable height tubes
  • Fluorescent colours available
  • Temperature resistance down to −196 °C