Urine and biological specimen containers, with ribbed cap
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
408001.1 408001.O 408001
DELT408001.1EA 142 EUR
DELT408001.1 DELT408001.O DELT408001
Urine and biological specimen containers, with ribbed cap
Diagnostische systemen Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
HDPE container and white ribbed cap.
  • Convenient handle for an easy single-handed use
  • Features a pharmaceutical grade joint, which eliminates the need for an insert plug, allows a better handling and provides a water tight seal
  • Containers are graduated in 50 ml increments up to 2 L
Ideal for 24 hour urine collection and any other liquid samples.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904