Biopsy processing cassettes, Histosette® II
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
445141 445147 445140 445143
631-9655EA 505 EUR
631-9655 631-9656 631-9657 631-9659
Biopsy processing cassettes, Histosette® II
Histologie-cassettes Cassettes voor biopsiebehandeling
Made of acetal polymer.1 mm square openings to maximise fluid exchange and to ensure proper drainage.
  • With 45° angle writing area
  • Suitable for automatic labellers
  • No need to break the lid and then put it in place, as it is supplied pre-mounted
  • Can be easily opened and closed with just one hand