Tissue processing cassettes, Histosette I
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
454141 454140 454143
DELT454141EA 125 EUR
DELT454141 DELT454140 DELT454143
Tissue processing cassettes, Histosette I
Histologie-cassettes Cassettes voor weefselbehandeling
Made of acetal polymer. With one-piece integral lid. To lock the cassette, just snap apart the lid, turn it round and lock it into the base of the cassette.
  • Writing area at 30° angle