Wire baskets, square
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H-210 H-240 H-200
216-7510EA 217 EUR
216-7510 216-7512 DELTH-200
Wire baskets, square
Manden Draadmanden
Square baskets, made of stainless steel wire, with folding handles.
  • Features a strong woven mesh base
  • Good chemical resistance
  • Good temperature resistance
  • Comes without lid (to order baskets with lid please contact our customer service