PCR manual workstation
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-540 M-542 M-541
212-9154EA 135 EUR
212-9154 212-9156 212-9216
PCR manual workstation
Rekken Tube Racks
This storage/working rack, with lid, is designed to work with different PCR volumes at the same time. It can accommodate up to 32 individual 0,2 ml PCR tubes, 24× 1,5/2,0 ml and 16× 0,5 ml microtubes.
  • Tube wells are alphanumerically matrixed with raised bold type for accurate sample identification
  • Rack has a transparent lid for easy viewing of samples
  • Frosted writing panels on four sides allow labelling or writing
  • Supplied with a removable rack accommodating 0,2 ml PCR tubes
Main rack W×D×H: 215×118×50 mm
Removable rack W×D×H: 125×85×15 mm