Centrifuge tubes, conical
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
401201 301212 301202
212-3362EA 46.6 EUR
212-3362 DELT301212 DELT301202
Centrifuge tubes, conical
Buisjes Centrifugebuisjes
PP or PS.
  • Good chemical resistance
  • Good temperature resistance
  • Conical bottom, with or without rim
Centrifugation:
PS tubes: 7,500 xg
PP tubes: 15,000 xg
Except code 301212: 3,000 xg