Face shield
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9725.514 9726.014
111-8231EA 55 EUR
111-8231 UVEX9726.014
Face shield
Gelaatsschermen
These polycarbonate and cellulose acetate visors can be attached to helmets by using a length adapter and can be locked in three positions.
  • Robust visor with specially developed adapter for attachment to side helmet slots
  • Extremely lightweight plastic design
  • Material thickness: 1 mm
Bestelinformatie: Visors are supplied with adapter but without helmet. Visors are compatible with helmets uvex airwing.