Falling ball viscometer
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
616-3282EA 0 EUR
616-3282
Falling ball viscometer
Viscosimeters Kogelvaltype viscometers
This falling ball viscometer provides an accurate way of measuring the viscosity, in transparent Newtonian liquids and gases.
Specially suited for the chemical (polymer solutions, solvents, inks), pharmaceutical (raw materials, glycerine), food (gelatin, sugar solutions) and mineral oil (oils, liquid hydrocarbons) industries.
Certificaten: DIN 53015, ISO 12058
Leveringsinformatie: Supplied with six balls, instrument case, thermometer, cleaning tools, calibration sheet and instruction manual.