96-well receiver plate, HTS Transwell®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3382 3583 3383 3783
734-4076EA 176.04 EUR
734-4076 734-4114 734-4077 734-2689
96-well receiver plate, HTS Transwell®
Microplaten Insertplaten en inserts
The HTS Transwell-96 permeable support has an array of 96-wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
  • Choice of either polyester (PET) membrane (0,4 µm, 1,0 µm, and 8,0 µm pore sizes) or polycarbonate (PC) membrane (0,4 µm, 3,0 µm, and 5,0 µm pore sizes)
  • 0,143 cm² membrane area per well, providing 20% to 50% more surface area for cell growth than other commercially available systems
  • Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access
  • Optimised for automation, with multichannel feeder ports, improved gripping surface, and standard bar codes
  • The reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabiliser to reduce the risk of spills during handling
  • The receiver plate isolates each well to enable 96 individual assays
  • Sterile
The HTS Transwell® 24-well permeable support has an array of 24 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 24 Transwell® supports to be handed in a single unit.
HTS Transwell® 96-well permeable support systems and plates The HTS Transwell® 96-well permeable support has an array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit. - Choice of either polyester (PET) membrane (1,0 µm or 8,0 µm pore sizes) or PC membrane (0,4 µm, 3,0 µm or 5,0 µm pore sizes) - Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access - Optimised for automation, with multichannel feeder ports, improved gripping surface, and standard barcodes - Reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabiliser to reduce the risk of spills during handling - Receiver plate isolates each well to enable 96 individual assays
The HTS Transwell® 96-well permeable support has an array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
Verpakking: The HTS Transwell-96 systems (0,4 µm PC, 0,4 µm PET, and 1,0 µm PET) are packaged with the 96-well insert plate in a reservoir plate and includes the 96-well receiver plate with lid. The HTS Transwell-96 plates (3,0 and 5,0 µm PC, 8,0 µm PET) are packaged with the 96-well insert plate in the 96-well receiver plate with lid. Reservoir plates may be purchased separately.