Test fluids for viscometers
Leverancier: HAAKE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
082-5042 082-5043 082-5044 082-5046 082-5336 082-5377 082-5335 082-5410 082-5412 082-5411
HAAK082-5042EA 0 EUR
HAAK082-5042 HAAK082-5043 HAAK082-5044 HAAK082-5046 HAAK082-5336 HAAK082-5377 HAAK082-5335 HAAK082-5410 HAAK082-5412 HAAK082-5411
Test fluids for viscometers
Standaarden Standaarden voor viscometrie
Test and certificated fluids for viscometery - mesurement temperature at 20 °C unless indicated.
Opgelet: * Measurement temperature 25 °C