Ultimate union kits and fittings
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G2855-20530 G3182-61581 G3182-61580 G2855-20555 G2855-20590 G2855-60200 G2855-60570 G3184-60065 G2855-20532 G3182-60581 5190-6989 5190-6990 5190-6993 G3182-20580
HEWLG2855-20530EA 39 EUR
HEWLG2855-20530 548-0709 548-0708 HEWLG2855-20555 HEWLG2855-20590 HEWLG2855-60200 HEWLG2855-60570 HEWLG3184-60065 HEWLG2855-20532 HEWLG3182-60581 HEWL5190-6989 HEWL5190-6990 HEWL5190-6993 HEWLG3182-20580
Ultimate union kits and fittings
Fittingen en connectoren
Ultimate union provides low dead volume column connections that are used to create an inert, leak-free, and robust seal.
  • No need to re-tighten after temperature cycles
Bestelinformatie: Kit includes union (deactivated or non-deactivated), internal nuts (2), swaging nut, and an oven wall clip.