Cell culture media, Ham's F12 (F-12 Nutrient Medium)
Leverancier: BIOWEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P0134-N1L P0134-N10L L0135-500 L0136-500 P0134-N5L
P0134-N1LEA 33 EUR
P0134-N1L P0134-N10L L0135-500 L0136-500 P0134-N5L
Cell culture media, Ham's F12 (F-12 Nutrient Medium)
Celcultuurmedia
Ham's F12 was originally developed for the serum-free clonal growth of Chinese Hamster Ovary (CHO) cells, lung cells and mouse L-cells. It is frequently used with dialysed serum, hormones, selenium and other supplements for serum-free cultures. It is the medium of choice for supporting the growth of cells of rodent origin, particularly rabbit and rat and has proved to be an excellent cloning medium for the culture of myeloma and hybrid cells.