Motoraangedreven luchtzuiverende maskers, Airvisor 2
Leverancier: HONEYWELL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1013939 1013980
DALL1013939EA 307.26 EUR
DALL1013939 DALL1013980
Motoraangedreven luchtzuiverende maskers, Airvisor 2
Ademhalingsmaskers Ademhalingssystemen
Airvisor 2 MV Chemical Kit with acetate chemical resistant visor, two disposable visor covers, fabric hood and neck cover, waist belt complete, carry bag. For applications exposed to chemicals.
Airvisor 2 MV Industry Kit with polycarbonate impact resistant visor, one fabric cassette, two disposable visor covers and head cover, waist belt complete, carrying bag. For industrial applications.
  • Lightweight product with wide field of vision
  • Hygienical single use head and neck protection
  • Visor keeps its original appearance due to visor cover
  • Carbon filter element removes odours
  • Hose food grade PVC material, complete with High Flow Quick Release connectors
Toevoerdruk 4,2 (58 psi) tot 7 bar (102 psi)
Debiet 290 l/min
Certificaten: EN 14594, EC Cat 3