Fused silica tubing cutter
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5183-4620 420-1000
HEWL5183-4620EA 543 EUR
HEWL5183-4620 HEWL420-1000
Fused silica tubing cutter
Snijders
Cutting tool, for fused silica tubing, with rotating diamond blade.