Pharma-zonemonsternemers, Novartos Uno
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5358-0140 5358-1001 5358-0120 5358-1400 5358-0110 5358-0600 5358-0105 5358-0115 5358-0125 5358-0130 5358-0135 5358-0103 5358-0100 5358-0630 5358-1030 5358-1430
300-0124EA 141 EUR
300-0124 300-0127 300-0122 300-0128 300-0120 300-0125 300-0119 300-0121 300-0123 300-0201 BURK5358-0135 BURK5358-0103 BURK5358-0100 331-0377 331-0378 331-0379
Pharma-zonemonsternemers, Novartos Uno
Monsternemers Monsternemers voor vaste stoffen
Novartos Uno sampler with two or three sample chambers, for one sample zone.
  • Novartos Uno for targeted sampling
  • The sample inserts fit precisely and can easily be replaced
  • Inserts are available in volumes from 0 to 4,0 ml (alternative sample volumes in increments of 0,1 ml on request)
  • The insertion depths can easily be read from the laser-engraved depth scale
  • Stainless steel AISI 316L (1.4404)/AISI 316Ti (1.4571)
  • Groove-free and electro-polished
  • 25 mm Ø
Novartos the Pharma zone sampler for sampling according to FDA guidelines for Unit-Dose-Sampling. It is now possible to withdraw a double- or triple-sample without inserting the lance repeatedly into the bulk material. Mixing is mainly prevented in this way. One significant advantage of Novartos is the easy cleaning. Complete filling and emptying of sample chambers has been improved substantially using computer-optimised geometry. There are no crevices or dead spaces where sampling residues can collect.