Flasks, flat bottom with standard ground joint
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0752-29-0500 0752-29-0100
NEUB0752-29-0500EA 132 EUR
NEUB0752-29-0500 NEUB0752-29-0100
Flasks, flat bottom with standard ground joint
Kolven Platbodemkolven
Borosilicate glass 3,3, amber.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Amber stained glass provides protection for materials sensitive to light.