Gown and hood dispenser
Leverancier: Palbam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113-8175EA 1170 EUR
113-8175
Gown and hood dispenser
PBM-dispensers
Stainless steel 304, electropolished finish wall-mounted dispenser for plastic wrapped gowns and hoods.
  • Easy refilling and easy retrieval
  • Designed to prevent spillage of items
  • Can be adapted for use as a general dispenser
For use in cleanrooms.
Certificaten: ISO certified
Leveringsinformatie: Typically available from stock for direct shipment from the supplier.