Dispensers with sloping top
Leverancier: Palbam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GD-2060-S GD-6060-S GD-4060-S GD-8060-S
113-8178EA 792 EUR
113-8178 113-8180 113-8179 113-8181
Dispensers with sloping top
PBM-dispensers
Stainless steel 304 with electropolished finish wall-mounted dispensers designed for the clean and efficient dispensing and storage of gloves, hairnets, face masks and shoe covers.
  • Eliminate particle build-up
  • Smooth curved opening for easy snag-free pull out
  • Sloping tops prevents articles being placed in dispensers
Suitable for applications in the cleanroom industries, such as pharmaceutical, biotechnology, medical device, microelectronics and nanotechnology.
Certificaten: ISO certified
Leveringsinformatie: Typically available from stock for direct shipment from the supplier.