Microtube racks, incubation, round
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-026 M-024
DELTM-026EA 228 EUR
DELTM-026 DELTM-024
Microtube racks, incubation, round
Rekken Tube Racks
PP
Even when the rack is fully loaded, tubes are completely immersed while tops rest above the rack, floating in a beaker of water. Perfect for use in benchtop incubation. The rack is easily unloaded by lightly pressing it down against the bench until the legs touch, releasing the tubes. Racks are 6,4 mm thick with 19 mm long legs and a central handle.