PCR tubes, 0,2 ml, with cap
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4094.1R 4094.1N
DELT4094.1REA 227 EUR
DELT4094.1R DELT4094.1N
PCR tubes, 0,2 ml, with cap
Buisjes PCR-buisjes
Tubes made of PP with attached hinged caps.
  • Caps are flat and easily pierceable
  • RNAse, DNAse and PCR inhibitors free