Solid phase extraction cartridges, Sep-Pak® Plus
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WAT036810 WAT020520 WAT020515
WATEWAT036810EA 261 EUR
WATEWAT036810 WATEWAT020520 WATEWAT020515
Solid phase extraction cartridges, Sep-Pak® Plus
Kolommen voor chromatografie
These cartridges can used with liquid and gas samples and have the unique capability of reverse flow direction, a useful feature for trace enrichment application.
  • Precision design suits to robotic application also more convenient in manual use
  • Sorbents are in short and long body configurations
Can be connected to different devices like filter units, positive flow pumps and syringes, vacuum manifolds, automated sample processor, and in-line reaction systems (PET applications).
Order Now
Specificaties
Environmentally Preferable