Solid phase extraction cartridges, Sep-Pak® Vac
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WAT043345 WAT036905 WAT020805 WAT054945 WAT043395 WAT054925 WAT054515 WAT020840 WAT020810
WATEWAT043345EA 0 EUR
WATEWAT043345 WATEWAT036905 WATEWAT020805 WATEWAT054945 WATEWAT043395 WATEWAT054925 WATEWAT054515 WATEWAT020840 WATEWAT020810
Solid phase extraction cartridges, Sep-Pak® Vac
Kolommen voor chromatografie
These cartridges deliver the option of minimum sample preparation. Consists of moulded, high purity PP syringe-barrel bodies with two PE frits to contain the packing material.
  • Standard male Luer permits use with multi-position vacuum manifolds
  • Colour coded printing on syringe barrels for identification