Trays, large deep
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19604 19603
DELT19604EA 134 EUR
DELT19604 DELT19603
Trays, large deep
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Trays made of natural HDPE.
  • Stackable
  • Good temperature resistance
  • Good chemical resistance
With ergonomically designed handles.