Wash bottles, narrow neck
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
44310000 44300000
DELT44310000EA 256 EUR
DELT44310000 DELT44300000
Wash bottles, narrow neck
Flessen Spuitflessen
HDPE, translucent, with blue HDPE screw cap, delivery tube and seal cap, natural.
  • High resistance to all liquids, acids and bases
  • Wide neck for safe and easy filling
  • Caps ribbed for a better handling