Scintillation vials
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
900100 900101
DELT900100EA 453 EUR
DELT900100 DELT900101
Scintillation vials
flesjes
Single use vials made of high density polyethylene to minimise solvent losses.
  • Screw cap ensures leakproof handling
  • Compatible with most liquid scintillation counters