Serum replacement, FreeAdd 1X
Leverancier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S6010-050 S6010-100 S6010-500
S6010-050EA 0 EUR
S6010-050 S6010-100 S6010-500
Serum replacement, FreeAdd 1X
Serum
FreeAdd 1X is a fully chemically defined substitute for animal serum. It provides the necessary nutritional support for cell growth, development and expression. It has been tested on a variety of cells and can be used in combination with basal medium to generate equal or even better cell culture performance.
  • No hydrolysates or mixtures
  • Non detectable endotoxin level
  • Broad range (cell lines, primary cells, stem cells)
  • GMP produced
  • 100% batch-to-batch consistency