Analytical HPLC columns, Symmetry® C8
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WAT046970 WAT054270 WAT058961 WAT066204 WAT045995
WATEWAT046970EA 865 EUR
WATEWAT046970 WATEWAT054270 WATEWAT058961 WATEWAT066204 WATEWAT045995
Analytical HPLC columns, Symmetry® C8
Kolommen voor chromatografie
Symmetry® C8 columns feature monofunctionally bonded C8 ligands on a high purity base-deactivated silica.
Symmetry® columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.