Analytical HPLC columns, Nova-Pak® C18
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WAT044380 WAT015220 WAT086344 WAT011695 WAT044375
WATEWAT044380EA 285 EUR
WATEWAT044380 WATEWAT015220 WATEWAT086344 WATEWAT011695 WATEWAT044375
Analytical HPLC columns, Nova-Pak® C18
Kolommen voor chromatografie
Nova-Pak® C18 columns are general purpose, silica based, reversed phase C18 columns that are based on 4 µm particle technology.
  • High resolution
  • Efficient separations
  • Fast chromatography