Analytical HPLC columns, SunFire™ C18
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
186002530 186003401 186002552 186002559 186002554 186002684 186002682 186003417 186002553 186002544 186002683
WATE186002530EA 0 EUR
WATE186002530 WATE186003401 WATE186002552 WATE186002559 WATE186002554 WATE186002684 WATE186002682 WATE186003417 WATE186002553 WATE186002544 WATE186002683
Analytical HPLC columns, SunFire™ C18
Kolommen voor chromatografie
SunFire™ columns provide symmetrical peak shapes for improved resolution. With new bonding and endcapping technologies, they offer great efficiency for acid, neutral, and basic compounds at low and intermediate pH ranges. They are compatible with mass spectrometry applications, providing sharp peaks, high sensitivity, large peak capacities and very low bleed.