Disposal cabinet
Leverancier: Palbam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PALBGDC8040-FRAMEEA 0 EUR
PALBGDC8040-FRAME 134-0486 134-0487
Disposal cabinet
Bakken Afvalbakken
This disposal cabinet can be used as a stand-alone unit or in combination with the dispenser unit (connecting frame required).
  • Made of stainless steel 304 with electropolished finish
  • Two waste openings, two doors and levelling feet
  • Does not require connecting to the wall
Can be used in cleanroom and critical environments.
Leveringsinformatie: Supplied with two large plastic bins.
Certificaten: ISO certified.
Leveringsinformatie: Typically available from stock for direct shipment from the supplier. Supplied with two large plastic bins.