Celzeven voor pipettips van 1000 µl, Flowmi™
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H136800040 H136800070
734-5950EA 279 EUR
734-5950 734-2709
Celzeven voor pipettips van 1000 µl, Flowmi™
Celzeven
Flowmi™-celzeven (poriegrootte van 40 µm) zorgen voor snelle, efficiënte filtering van monsters in kleine volumes (tot 1000 µl) en een ongeëvenaarde opslag van monstervolumes.
  • Preserves volume when filtering small sample volumes
  • Removes cell aggregates and creates uniform single cell suspensions
  • Decreases the potential for clogging of FLOW or FACS instruments by effectively filtering cellular debris
  • Sterile: 40 or 70 µm porosity
Flowmi™ is recommended for use with samples having a maximum concentration of 2 million cells/ml (40 µm) or 4 million cells/More Product Information
Order Now
Specificaties
Environmentally Preferable