Online SPE products, HyperSep™ Retain™ PEP
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60310-201 60310-202 60311-201 60311-202 60312-201 60312-202
HYPE60310-201EA 484 EUR
HYPE60310-201 HYPE60310-202 HYPE60311-201 HYPE60311-202 HYPE60312-201 HYPE60312-202
Online SPE products, HyperSep™ Retain™ PEP
Kolommen voor chromatografie
These online SPE products are designed to clean-up and concentrate target analytes.
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds