Deep freeze boxes, Rotilabo®
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A272.1 A273.1 A274.1 A275.1 1597.1 1598.1 1599.1
ROTHA272.1EA 24.1 EUR
ROTHA272.1 ROTHA273.1 ROTHA274.1 ROTHA275.1 ROTH1597.1 ROTH1598.1 ROTH1599.1
Deep freeze boxes, Rotilabo®
Dozen Koelboxen en koeltassen
HDPE box with LDPE lid.
  • Suitable for foodstuffs
  • Stackable
  • With notched lid
  • Rectangular and slightly conical
  • Temperature stable from −40 to +95 °C