Online SPE products, HyperSep™ Retain™ AX
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60310-401 60310-402 60311-401 60311-402 60312-401 60312-402
HYPE60310-401EA 507 EUR
HYPE60310-401 HYPE60310-402 HYPE60311-401 HYPE60311-402 HYPE60312-401 HYPE60312-402
Online SPE products, HyperSep™ Retain™ AX
Kolommen voor chromatografie
HyperSep™ Retain™-AX for retention of acidic and non-polar analytes in a sample matrix when used with an appropriate HPLC column.
  • Effective removal of contaminants such as proteins from samples
  • Compatible with conventional HPLC systems
  • Fast and effective clean-up and concentration of target compounds