Disposal Boxes for Broken Glasses
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KN69.1 KN70.1
ROTHKN69.1EA 118.5 EUR
ROTHKN69.1 ROTHKN70.1
Disposal Boxes for Broken Glasses
Dozen Afvaldozen
Disposal boxes are made of cardboard, with 50 μm thick polyethylene liner bag, which prevents leakage of liquid residues.
  • Large flap in lid for easy filling
  • Flat folded for space-saving storage
  • Printed with eye-catching graphics in four languages (English, German, French, Spanish)
Ideal for safe collection and disposal of defective glassware.