HPLC columns for online SPE, HyperSep™ Hypercarb™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60310-501 60310-502 60311-501 60311-502 60312-501 60312-502
HYPE60310-501EA 543 EUR
HYPE60310-501 HYPE60310-502 HYPE60311-501 HYPE60311-502 HYPE60312-501 HYPE60312-502
HPLC columns for online SPE, HyperSep™ Hypercarb™
Kolommen voor chromatografie
When used with conventional HPLC systems and appropriate switching valves, these columns provide effective removal of contaminants, such as proteins, from samples and fast, effective clean-up and concentration of target compounds.