Sample loops for Valco valves
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-SLV-76-10 JR-SLV-76-100 JR-SLV-76-1000 JR-SLV-76-20 JR-SLV-76-250 JR-SLV-76-5 JR-SLV-76-50 JR-SLV-76-500
JOURJR-SLV-76-10EA 94.7 EUR
JOURJR-SLV-76-10 JOURJR-SLV-76-100 JOURJR-SLV-76-1000 JOURJR-SLV-76-20 JOURJR-SLV-76-250 JOURJR-SLV-76-5 JOURJR-SLV-76-50 JOURJR-SLV-76-500
Sample loops for Valco valves
Benodigdheden voor chromatografie Injectieloops voor chromatografiesystemen
These VICI Jour sample loops are made of high quality stainless steel 316.
  • Fits all Valco W and UW type valves
  • High pressure resistance, low permeability
  • Tolerance of ±0,05 mm and pressure rating of <350 bar
They are cut using an electrolytic cutting process, cleaned with ultrasound and high pressure steam to remove contaminants, dried, and capped with softener-free caps to avoid recontamination of the tubing.
Leveringsinformatie: Supplied with stainless hex-head nuts and stainless ferrules for reliable connections.
Order Now
Specificaties
Environmentally Preferable