ESD occupational footwear, clogs, ALPRO by Birkenstock®, C100
Leverancier: LIEFERSERVICE UND LOGISTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
061418/36 061400/44 061400/45 061400/46 061400/47 061400/48 061400/39 061400/40 061400/41 061400/42 061400/43 061418/37 061418/38 061418/39 061418/40 061418/41 061418/42 061410/39 061410/45 061410/46 061410/47 061410/48 061410/40 061410/41 061410/42 061410/43 061410/44 061408/36 061408/37 061408/38 061408/39 061408/40 061408/41 061408/42 061390/46 061398/42 061398/41 061398/40 061398/39 061398/38 061398/37 061398/36 061390/47 061390/45 061390/44 061390/43 061390/42 061390/41 061390/40 061390/39 061390/48
114-4779EA 109 EUR
114-4779 114-6179 114-6180 114-6181 114-6182 114-6183 114-6174 114-6175 114-6176 114-6177 114-6178 114-4780 114-4781 114-4782 114-4783 114-4784 114-4785 114-4769 114-4775 114-4776 114-4777 114-4778 114-4770 114-4771 114-4772 114-4773 114-4774 114-6184 114-6185 114-6186 114-6187 114-6188 114-6189 114-6190 114-4759 114-4768 114-4767 114-4766 114-4765 114-4764 114-4763 114-4762 114-4760 114-4758 114-4757 114-4756 114-4755 114-4754 114-4753 114-4752 114-4761
ESD occupational footwear, clogs, ALPRO by Birkenstock®, C100
Schoenen en laarzen
Breathable ESD clogs made from smooth leather with cork–Latex insole and anti-slip outer sole.
  • Resistance range from 105 to 3,4×107 ohm
  • Anti-slip, abrasion resistant sole (compliant with DIN 4843 part 100)
  • Adjustable heel strap and instep strap
  • Anatomical shape and deep heel cup
  • Cushioning cork sole
For all areas where increased protection against electrostatic discharge is required.
Colour: White, blue or black
Certificaten: EN ISO 61340