Caps for solvent safety bottles, Q-Series
Leverancier: OMNIFIT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00932Q-3V 00938Q-2 00938Q-3 00945Q-3 00945Q-4 00945Q-4V
OMNI00932Q-3VEA 342 EUR
OMNI00932Q-3V OMNI00938Q-2 OMNI00938Q-3 OMNI00945Q-3 OMNI00945Q-4 OMNI00945Q-4V
Caps for solvent safety bottles, Q-Series
Sluitingen Flesdoppen
Outer PP cap with PTFE inner body.
  • Optional valves offer easy on-off control of individual fluid lines, ideal for selecting flow or for closing off lines prior to removing caps to change or re-fill solvent bottles
  • Robust threaded ports for standard ¹/₄"-28 UNF threaded fitting nuts and adaptors give a secure connection ideal for repeated connect / disconnect
  • Optional screw-in check valve and filter accessory enables pressure equalization while keeping the system closed and preventing both particulate contamination and evaporation of expensive or harmful