Nitrogen and Sulphur chemiluminescence detectors supplies
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G1530-80650 G6600-60037 G6600-60038 G6600-67007 G6600-67008 G6600-80042 G6600-80043 G6600-80044 G6600-80045 G6600-80050 G6600-80051 G6600-80063 G6600-85000 G6600-85001 G6600-85002 G2630-60710 G2933-85001 G6600-60035 G6600-80018
HEWLG1530-80650EA 389 EUR
HEWLG1530-80650 HEWLG6600-60037 HEWLG6600-60038 HEWLG6600-67007 HEWLG6600-67008 HEWLG6600-80042 HEWLG6600-80043 HEWLG6600-80044 HEWLG6600-80045 HEWLG6600-80050 HEWLG6600-80051 HEWLG6600-80063 HEWLG6600-85000 HEWLG6600-85001 HEWLG6600-85002 HEWLG2630-60710 HEWLG2933-85001 HEWLG6600-60035 HEWLG6600-80018
Nitrogen and Sulphur chemiluminescence detectors supplies
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Nitrogen Chemiluminescence Detector (NCD) is a nitrogen-specific detector that produces a linear and equimolar response to nitrogen compounds based on a chemiluminescent reaction of NO with ozone.
Sulphur Chemiluminescence Detector (SCD) for the analysis of sulphur compounds.