Ice buckets, Bel-Art Magic Touch 2™
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M168072004 M168072003 M168072002 M168072001 M168074003 M168074001 M168074004 M168074002
216-1084EA 284 EUR
216-1084 216-1083 216-1082 216-1081 216-1087 216-1085 216-1088 216-1086
Ice buckets, Bel-Art Magic Touch 2™
IJscontainers
Expanded urethane
  • Impervious to moisture and odours
  • Temperature range: −196 to 100 °C
  • Features convenient pour spout
  • Snug-fitting, keyed lid and base make stacking easy and secure
Magic Touch 2™ laboratory ice buckets are lightweight, durable and non-sweating. Superior insulating properties combined with high chemical resistance make them compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, icesalt slurries, dry ice solvent slurries, or liquid nitrogen.
Bestelinformatie: Magic Touch 2™ ice buckets are available in 2,5 and 4,0 L sizes, each in four colors; black, blue, cherry red and emerald green. They either come as individually packaged buckets or in bulk quantities of 12.