Capillary columns, TraceGOLD™ TG-624SilMS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26059-4950 26059-3330 26059-3390 26059-3410 26059-3960 26059-4080 26059-3320
555-8781EA 617 EUR
555-8781 555-8788 555-8787 555-8786 555-8785 555-8784 555-8789
Capillary columns, TraceGOLD™ TG-624SilMS
Kolommen voor chromatografie
High thermal stability column ideal for volatile organic analysis.
  • High thermal stability - maximum temperatures up to 320 °C
  • Highly inert - excellent peak shape for a wide range of compounds