Capillary columns, TraceGOLD™ TG-ALC Plus I and TG-ALC Plus II
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26063-2240 26063-2980 26063-3390 26063-3960
555-8780EA 874 EUR
555-8780 555-8779 555-8778 555-8777
Capillary columns, TraceGOLD™ TG-ALC Plus I and TG-ALC Plus II
Kolommen voor chromatografie
Application-specific columns for blood alcohol analysis.
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
  • Fast analysis time giving full separation of blood alcohols