Chemical resistant overalls, Body Plus
Leverancier: BEST FRANCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01073A010XL 01073A010XXL
BESF01073A010XLEA 0 EUR
BESF01073A010XL BESF01073A010XXL
Chemical resistant overalls, Body Plus
Wegwerpkleding
These hooded overalls are made of breathable SMS fabric. They offer protection against hazardous dust and light liquid chemical splashes and sprays.
  • Elastic wrists, ankles and back
  • Moisture wicking properties
  • Zip closure with adhesive flap cover
Colour: White
Weight: 50 g/m²
Certificaten: Type 5/6; Cat. III