O-ring seals for flat flanges, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PS100 PS75
BIBBPS100EA 39.5 EUR
BIBBPS100 BIBBPS75
O-ring seals for flat flanges, Quickfit®
Afdichtingen Afdichtringen
PTFE, white.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Forms tight seal with flat flange joints
For use with Quickfit® flat flange jointed glassware.