Cleaning cloths, Tork, W8
Leverancier: Essity
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Tork®
115-3121EA 104 EUR
115-3121
Cleaning cloths, Tork, W8
Schoonmaakdoekjes Herbruikbare reinigingsdoeken Doeken voor algemeen gebruik
This soft and flexible multipurpose cloth made of chemical pulp, polyester and PP quickly absorbs liquids and oil. The cloth provides a professional cleaning result thanks to the exelCLEAN™ effect.
  • Long-lasting cloth suitable for repeated use in wet & dry conditions
  • Efficient absorb and release properties mean less product needed to do the same job
  • Single wipe dispensing in portable and small size format for compact spaces
Food contact approved.