Nitril handschoenen, Peha-soft® Nitrile PF
Leverancier: HARTMANN PAUL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9421912 9421902 9421922 9421892 9421932
HART9421912EA 24.3 EUR
HART9421912 HART9421902 HART9421922 HART9421892 HART9421932
Nitril handschoenen, Peha-soft® Nitrile PF
Handschoenen
These reversible disposable gloves are made of nitrile rubber, a synthetic material which provides all the advantages of latex without causing allergic reactions as commonly found with natural latex.
  • Resistant to germs, bacteria, viruses and chemicals
  • Powder-free, therefore eliminating danger of food contamination due to corn starch
  • Resistant to a large number of chemicals, such as disinfectants
  • Optimal grip and polymeric internal coating for good tactility
The gloves are suitable for a range of uses within the food processing industry. They provide very high protection against bacterial hazards and are therefore excellent for use in the laboratory. The Peha-soft nitrile pf gloves can also be used when disinfecting bottles.
Length: 240 mm
Thickness: 0.07 mm (sleeve); 0.10 mm (palm); 0.10 mm (fingertip)
Colour: blue
EN 374, EN 455; Cat. III; AQL 1.5
Certificaten: EN 374, EN 455; Cat. III; AQL 1,5
Goedgekeurd voor voedselcontact